Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
PIG® Absorbent pans

PIG® Absorbent pans

Pans, 33,6 L, 270×270×80 mm
N° de catalogue NEWP2410PP-01
Fournisseur:   NEW PIG
PIG® Absorbent pans
N° de catalogue NEWP2410PP-01
Fournisseur:  NEW PIG
PIG® Absorbent pans
PIG® Absorbent pans

Pans, 33,6 L, 270×270×80 mm

Prix
119,00 €
Connexion Pour la disponibilité
1 * 12 ST
Product Details & Documents

Trap annoying drips in this spillproof, absorbent-filled Drip Pan. Polypropylene cover prevents splashes and keeps filler from spilling out.

  • Low-profile drip pan fits underneath drum taps and other tight spaces; wide base won't tip over like unsturdy coffee cans
  • Soft polypropylene cover catches drips without splashing and prevents highly absorbent polypropylene filler from spilling out
  • Highly absorbent polypropylene filler soaks up liquid on contact
  • Ideal for use under leaking drum taps, spigots and flanges to absorb nuisance drips; keeps your floors clean and safe
  • Absorbs and retains oils, coolants, solvents and water

98% pre-consumer recycled polypropylene filler.

Spécifications
  • Désignation:
    Pans
  • l×p×h:
    270×270×80 mm
  • Capacité d'absorption:
    33,6 L