PIG® Absorbent pans
Pans, 67,2 L, 270×270×80 mm
N° de catalogue NEWP2410PP
Fournisseur: NEW PIG
Aller à :
Product Details & Documents
Trap annoying drips in this spillproof, absorbent-filled Drip Pan. Polypropylene cover prevents splashes and keeps filler from spilling out.
- Low-profile drip pan fits underneath drum taps and other tight spaces; wide base won't tip over like unsturdy coffee cans
- Soft polypropylene cover catches drips without splashing and prevents highly absorbent polypropylene filler from spilling out
- Highly absorbent polypropylene filler soaks up liquid on contact
- Ideal for use under leaking drum taps, spigots and flanges to absorb nuisance drips; keeps your floors clean and safe
- Absorbs and retains oils, coolants, solvents and water
98% pre-consumer recycled polypropylene filler.
Spécifications
- Désignation:Pans
- l×p×h:270×270×80 mm
- Capacité d'absorption:67,2 L
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



