Disposable cannulas
Disposable cannulas, G19, crème, 120 mm, +, 1,1×120 mm
Fournisseur: EHRHARDT SOEHNE
Aller à :
Product Details & Documents
Disposable cannulas.
- Pyrogen-free, not toxic
- With lancet cut
- EO-sterilisation
Documents
Certificats
Rechercher par numéro de lotSpécifications
- Ø×L:1,1×120 mm
- Jauge:G19
- Stérile:+
- Couleur:crème
- Désignation:Disposable cannulas
- Longueur:120 mm
- Seq:29
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



