Blood collection needles, BD Vacutainer® Eclipse™
Blood collection needles, BD Vacutainer® Eclipse™
21, 32 mm, vert
Catalogus # BDAM368609
Leverancier:   BD Medical
Blood collection needles, BD Vacutainer® Eclipse™
Catalogus # BDAM368609
Leverancier:  BD Medical
Blood collection needles, BD Vacutainer® Eclipse™
Blood collection needles, BD Vacutainer® Eclipse™
21, 32 mm, vert
Price
30,70 €
Log in voor beschikbaarheid
1 * 48 ST
Product Details & Documents
The BD Vacutainer® Eclipse™ blood collection needle is a safety-engineered, multi-sample blood collection needle that offers a simple, effective way to collect blood while reducing the possibility of needlestick injuries. It features a safety shield that allows for one-handed activation to cover the needle immediately upon withdrawal from the vein and confirms proper activation with an audible click.
  • Built-in safety shield is aligned with the bevel of the needle
  • The safety shield permanently locks into place, significantly reducing the risk of needlestick injuries
  • The pre-attached holder (only supplied with some models) minimises the risk of accidental needlestick injuries from the holder end of the device
Specificaties
  • Code couleur:
    vert
  • Jauge:
    21
  • Longueur:
    32 mm
  • Seq:
    01
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.