Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Calibrated Baumé hydrometers

Calibrated Baumé hydrometers

–2 - 10, 0,1, 300 mm
N° de catalogue ALLA0101FC002/20QP
Fournisseur:   Alla France
Calibrated Baumé hydrometers
N° de catalogue ALLA0101FC002/20QP
Fournisseur:  Alla France
Calibrated Baumé hydrometers
Calibrated Baumé hydrometers

–2 - 10, 0,1, 300 mm

Prix
67,80 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Baumé hydrometers calibrated at 20 °C.

Spécifications
  • Longueur:
    300 mm
  • Seq:
    01
  • Division:
    0,1
  • Plage de mesure:
    –2 - 10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.