Eau, BAKER ANALYZED®, J.T.Baker®
Flacon Verre, 2,5 L
Fournisseur: Avantor
Eau, BAKER ANALYZED®, J.T.Baker®
N° de catalogue 9825.2500GL
Fournisseur: Avantor
Numéro CAS: 7732-18-5
Aller à :
Product Details & Documents
Synonymes: Hydrogen oxide
- Numéro CAS:7732-18-5
- Formule:H₂O
- Numéro MDL:MFCD00011332
- Poids moléculaire:18.02 g/mol
- Index Merck:13,10098
- Point d'ébullition:100 °C (1013 hPa)
- Point de fusion:0 °C
- Densité:1 g/cm³ (4 °C)
- Température de stockage:Température ambiante
- EINECS:231-791-2
Documents
Certificats
Rechercher par numéro de lotsds_BAKR9825
SDS
spec_BAKR9825.1000GL.pdf spec_BAKR9825.2500GL.pdf
Product specification
VWR J.T.Baker® High Purity solvents and reagents
Product Literature
Spécifications
Résultats des tests de spécification
- Residue after Evaporation:max. 5 ppm
- LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid::-
- at 210 nm:max. 0,005
- at 254 nm:max. 0,001 uv
- LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid::-
- Positive ESI-MS Sensitive Impurities (as Reserpine):max. 100 ng/ml
- Trace Impurities (in ppb):-
- Aluminium (Al):max. 500 ppb
- Calcium (Ca):max. 100 ppb
- Iron (Fe):max. 100 ppb
- Magnesium (Mg):max. 100 ppb
- Potassium (K):max. 100 ppb
- Sodium (Na):max. 100 ppb
- Product Information (not specifications)::-
- Density (g/ml) at 20°C:1.00
Spécifications
- Type de conditionnement:Flacon Verre
- Cdt:2,5 L
- Cat. No.:9825.2500GL
- Certification environnementale:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Safer Human and Environmental Health



