Inserts d'injection
Insert hélicoïdal, extrémité ouverte, désactivé
Catalog # HEWL5188-5396
Supplier:   Agilent
Inserts d'injection
Catalog # HEWL5188-5396
Supplier:  Agilent
Inserts d'injection
Inserts d'injection
Insert hélicoïdal, extrémité ouverte, désactivé
Price
122,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
Specifications
  • Désignation:
    Insert hélicoïdal, extrémité ouverte, désactivé
  • Seq:
    64