Percoll®, Siam
Percoll®, Siam
Percoll®
N° de catalogue P1644-1L
Fournisseur:   SIGMA ALDRICH MICROSCOPY
Percoll®, Siam
N° de catalogue P1644-1L
Percoll®, Siam
Percoll®
Prix
1 060,00 €
Connexion Pour la disponibilité
1 * 1 L
Product Details & Documents
Percoll® is a classic medium for density gradient centrifugation of cells, viruses, and subcellular particles. It consists of colloidal silica particles of 15 – 30 nm diameter (23% w/w in water), coated with polyvinylpyrrolidone (PVP).
  • Non toxic
  • Ideal for use with biological materials
Documents
Spécifications
  • Désignation:
    Percoll®
  • Seq:
    01