Pointes à filtre, aeroject®
Standard, 1 - 100 µl, +, Pipettes couramment utilisées, 10 portoirs, 96 cônes chacun
RATI1770100
: RATIOLAB
These sterile pipette tips with filters effectively prevent the pipettor shaft from contamination with aerosols and liquids.
- Free of RNase-, DNase-, DNA-, pyrogens, ATP
- Free of human DNA and PCR inhibitors
- Tips and aerosol filters made of ultra-hydrophobic PE-HD
- Minimised retention due to special surface treatment
- Perfect and tight fit on all major pipettor brands
- Tips with extended length: Ideal for operation with narrow vessels and reduced risk of cross-contamination
: Packed in plastic racks with hinged lids.
- Emballage:10 portoirs, 96 cônes chacun
- Stérile:+
- Volume:1 - 100 µl
- Style de pointe:Standard
- Pour l'utilisation avec:Pipettes couramment utilisées