Pointes à filtre, aeroject®
Standard, 1 - 100 µl, +, Pipettes couramment utilisées, 10 portoirs, 96 cônes chacun
  RATI1770100
 :   RATIOLAB
Pointes à filtre, aeroject®
  RATI1770100
 :  RATIOLAB
Pointes à filtre, aeroject®
Pointes à filtre, aeroject®
Standard, 1 - 100 µl, +, Pipettes couramment utilisées, 10 portoirs, 96 cônes chacun
128,00 €
1 * 960 ST
These sterile pipette tips with filters effectively prevent the pipettor shaft from contamination with aerosols and liquids.
  • Free of RNase-, DNase-, DNA-, pyrogens, ATP
  • Free of human DNA and PCR inhibitors
  • Tips and aerosol filters made of ultra-hydrophobic PE-HD
  • Minimised retention due to special surface treatment
  • Perfect and tight fit on all major pipettor brands
  • Tips with extended length: Ideal for operation with narrow vessels and reduced risk of cross-contamination
 : Packed in plastic racks with hinged lids.
  • Emballage:
    10 portoirs, 96 cônes chacun
  • Stérile:
    +
  • Volume:
    1 - 100 µl
  • Style de pointe:
    Standard
  • Pour l'utilisation avec:
    Pipettes couramment utilisées