Flacons à tare
30 ml, 50×25 mm
  HECH44251013
Flacons à tare
  HECH44251013
Flacons à tare
Flacons à tare
30 ml, 50×25 mm
21,50 €
1 * 1 ST
Made of borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
With interchangeable lid and frosted label.
  • Ø×h:
    50×25 mm
  • Capacité:
    30 ml