Flacons à tare
30 ml, 50×25 mm
HECH44251013
Made of borosilicate glass.
- Excellent chemical resistance
- Excellent temperature resistance
With interchangeable lid and frosted label.
- Ø×h:50×25 mm
- Capacité:30 ml
Made of borosilicate glass.
With interchangeable lid and frosted label.