Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Flacons à tare

Flacons à tare

70 ml, 40×80 mm
N° de catalogue HECH44250005
Flacons à tare
N° de catalogue HECH44250005
Flacons à tare
Flacons à tare

70 ml, 40×80 mm

Prix
27,90 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Made of borosilicate glass.

  • Excellent chemical resistance
  • Excellent temperature resistance

With interchangeable lid and frosted label.

Spécifications
  • Ø×h:
    40×80 mm
  • Capacité:
    70 ml
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.