Flasks, flat bottom with standard ground joint
2000 ml, 29/32 NS
N° de catalogue HECH41911036
Fournisseur: GLASWARENFABRIK KARL HECHT
Flasks, flat bottom with standard ground joint
N° de catalogue HECH41911036
Fournisseur: GLASWARENFABRIK KARL HECHT
Aller à :
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
Spécifications
- Rodage femelle:29/32 NS
- Capacité:2000 ml
- Seq:07