Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
1000 ml, 29/32 NS
N° de catalogue HECH41911030
Flasks, flat bottom with standard ground joint
N° de catalogue HECH41911030
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
1000 ml, 29/32 NS
Prix
56,70 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
Spécifications
  • Rodage femelle:
    29/32 NS
  • Capacité:
    1000 ml
  • Seq:
    06