Slide label rolls, polyester
Slide label, Polyester, blanc, 24×19 mm, On roll
Catalog # KLIN1513BR
Supplier: KLINIPATH
Jump to:
Product Details & Documents
These slide labels are made of polyester and are chemical resistant.
Specifications
- Couleur:blanc
- Désignation:Slide label
- Dimensions:24×19 mm
- Matière:Polyester
- Seq:003
- Emballage:On roll