Tourniquets
Tourniquet for babies, blue
N° de catalogue TERUXX-TCB2
Fournisseur: TERUMO
Aller à :
Product Details & Documents
These tourniquets are available in three sizes suitable for adults, children or babies.
- Sturdy, safety snap closure with one-hand operation
- Replaceable and washable ribbon
- Metal-, latex- and phthalate-free
Designed for daily use in all clinical settings including ambulance.
Documents
TERUXX-TC_TDS_EN
Fiche technique
Spécifications
- Désignation:Tourniquet for babies, blue
- Seq:03