Microplate stands
Microplate stands
Well plate stand, adjustable up to 45° angle
N° de catalogue DIVEWPAJ-1000
Fournisseur:   DIVERSIFIED BIOTECH
Microplate stands
N° de catalogue DIVEWPAJ-1000
Fournisseur:  DIVERSIFIED BIOTECH
Microplate stands
Well plate stand, adjustable up to 45° angle
Prix
293,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Convenient holder for microplates to comfortably view samples and pipette without strain. Available tilted at a 30° angle or as an adjustable version allowing the flexibility to adjust the angle of the microplate (45° max. tilt).
Spécifications
  • Désignation:
    Well plate stand, adjustable up to 45° angle
  • Seq:
    01