Reservoir system
Reservoir system
Frame for liquid reservoirs
N° de catalogue BECL372795
Marketsource
Fournisseur:   Beckman Coulter
Reservoir system
N° de catalogue BECL372795
Fournisseur:  Beckman Coulter
Marketsource
Reservoir system
Reservoir system
Frame for liquid reservoirs
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
This modular reservoir system holds liquids for pipetting with a manual pipettor or liquid handling systems including the full suite of Biomek liquid handlers.
  • Half and quarter reservoirs must be used with a frame
  • Full reservoirs hold up to 150 ml of liquid, and do not require a frame
  • Each frame can accommodate up to four quarter reservoirs or two half reservoirs
  • All reservoirs are sterile and made of PP
Not for use with 96 and 384 multi channel pipetting heads.
Documents
Spécifications
  • Désignation:
    Frame for liquid reservoirs
  • Seq:
    04