Reservoir system
Frame for liquid reservoirs
N° de catalogue BECL372795
Marketsource
Fournisseur: Beckman Coulter
Aller à :
Product Details & Documents
This modular reservoir system holds liquids for pipetting with a manual pipettor or liquid handling systems including the full suite of Biomek liquid handlers.
- Half and quarter reservoirs must be used with a frame
- Full reservoirs hold up to 150 ml of liquid, and do not require a frame
- Each frame can accommodate up to four quarter reservoirs or two half reservoirs
- All reservoirs are sterile and made of PP
Not for use with 96 and 384 multi channel pipetting heads.
Documents
Certificats
Rechercher par numéro de lotSpécifications
- Désignation:Frame for liquid reservoirs
- Seq:04