Stoppers, with drip tip, MBL®
Stoppers, with drip tip, MBL®
7/16, With pointed bottom
Catalog # BIBBTRF690
Supplier:   SciLabware
Stoppers, with drip tip, MBL®
Catalog # BIBBTRF690
Supplier:  SciLabware
Stoppers, with drip tip, MBL®
Stoppers, with drip tip, MBL®
7/16, With pointed bottom
Price
55,20 €
Log in for availability
1 * 10 ST
Product Details & Documents
Ideally suited for use with volumetric flasks etc. Lower closed end of stopper features a 'drip-tip' to ensure adhered liquid is returned to the flask. Flat head allows stopper to be stood on end, thus preventing contamination of the ground surface.
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Hollow-blown to reduce weight
Specifications
  • RN:
    7/16
  • Type:
    With pointed bottom
  • Seq:
    01