Stoppers, with drip tip, MBL®
7/16, With pointed bottom
Catalog # BIBBTRF690
Supplier: SciLabware
Jump to:
Product Details & Documents
Ideally suited for use with volumetric flasks etc. Lower closed end of stopper features a 'drip-tip' to ensure adhered liquid is returned to the flask. Flat head allows stopper to be stood on end, thus preventing contamination of the ground surface.
- Hexagonal head prevents the stopper from rolling and provides a good grip
- Hollow-blown to reduce weight
Specifications
- RN:7/16
- Type:With pointed bottom
- Seq:01