Trypsin Protease, HyClone™
Trypsin, 0,25% 1X, with 2,5 g porcine trypsin in HBSS with 0,2 g/EDTA, without calcium, magnesium (liquid), 500 mL
N° de catalogue HYCLSH30042.02
Trypsin Protease, HyClone™
N° de catalogue HYCLSH30042.02
Numéro CAS:  
Trypsin Protease, HyClone™
Trypsin Protease, HyClone™
Trypsin, 0,25% 1X, with 2,5 g porcine trypsin in HBSS with 0,2 g/EDTA, without calcium, magnesium (liquid), 500 mL
Numéro CAS:  
Prix
41,00 €
Connexion Pour la disponibilité
1 * 500 mL
Product Details & Documents
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium
Spécifications
Spécifications
  • Cdt:
    500 mL
  • Désignation:
    Trypsin, 0,25% 1X, with 2,5 g porcine trypsin in HBSS with 0,2 g/EDTA, without calcium, magnesium (liquid)
Spécifications