Cold traps, Type KF
Cold traps, Type KF
Cold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent cap
N° de catalogue KGWA17372
Fournisseur:   KGW
Cold traps, Type KF
N° de catalogue KGWA17372
Fournisseur:  KGW
Cold traps, Type KF
Cold traps, Type KF
Cold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent cap
Prix
643,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminium
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : [email protected]
Spécifications
  • Désignation:
    Cold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent cap
  • Seq:
    04
Accessories
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • KGWA1733
    Plastic ring for cold trap, type KF
    Connexion Pour la disponibilité
    1 * 1 ST - 62,50 €
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • KGWA1741
    Cooling finger S 29-GL
    Connexion Pour la disponibilité
    1 * 1 ST - 275,00 €