Biopsy specimen processing, CellSafe™
Biopad biopsy pad, blue
Catalog # 720-2331
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Tissu mousse Q Path bleu 1 * 500 Pce Catalog #720-2254
Product 720-2254 is similar to 720-2331 but may have a different manufacturer and/or packaging.
EPONGE A BIOPPSY CARRE BLEU 1 * 1.000 Pce Catalog #720-0211
Product 720-0211 is similar to 720-2331 but may have a different manufacturer and/or packaging.
Product Details & Documents
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
- No tissue damage
- Low reagent carry-over
The CellSafe™ fits in every embedding cassette.
Documents
Specifications
- Désignation:Biopad biopsy pad, blue