Eye showers
Eye shower, shower for faucet mounting
N° de catalogue 118-0797
Fournisseur: BRADLEY
Aller à :
Product Details & Documents
Eye shower for faucet mounting.These can be operated with a single movement that is the valve remains open automatically after activation.
- Pipe and fittings: brass, chromed
- Chromed spray heads, with dust caps and flow regulator
Delivery information: Included international pictogram board, assembly instruction and inspection label.
Spécifications
- Désignation:Eye shower, shower for faucet mounting
- Seq:01