Eye showers
Eye showers
Eye shower, shower for faucet mounting
N° de catalogue 118-0797
Fournisseur:   BRADLEY
Eye showers
N° de catalogue 118-0797
Fournisseur:  BRADLEY
Eye showers
Eye shower, shower for faucet mounting
Prix
131,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Eye shower for faucet mounting.These can be operated with a single movement that is the valve remains open automatically after activation.
  • Pipe and fittings: brass, chromed
  • Chromed spray heads, with dust caps and flow regulator
Delivery information: Included international pictogram board, assembly instruction and inspection label.
Spécifications
  • Désignation:
    Eye shower, shower for faucet mounting
  • Seq:
    01