Extraction thimbles
Extraction thimbles
22×60 mm, 2,0 mm
N° de catalogue 537-0043
Extraction thimbles
N° de catalogue 537-0043
Extraction thimbles
Extraction thimbles
22×60 mm, 2,0 mm
Prix
165,00 €
Connexion Pour la disponibilité
1 * 25 ST
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Spécifications
  • Ø int.×h:
    22×60 mm
  • Seq:
    24
  • Épaisseur:
    2,0 mm