Trypsin Protease, HyClone™
Trypsin Protease, HyClone™
Trypsin, 0,05% 1X, with 0,5 g porcine trypsin in HBSS with 0,2 g/l EDTA, without calcium, magnesium (liquid), 100 ml
N° de catalogue HYCLSH30236.01
Trypsin Protease, HyClone™
N° de catalogue HYCLSH30236.01
Numéro CAS:  
Trypsin Protease, HyClone™
Trypsin Protease, HyClone™
Trypsin, 0,05% 1X, with 0,5 g porcine trypsin in HBSS with 0,2 g/l EDTA, without calcium, magnesium (liquid), 100 ml
Numéro CAS:  
Prix
15,00 €
Connexion Pour la disponibilité
1 * 100 mL
Product Details & Documents
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium
Documents
Spécifications
Spécifications
  • Cdt:
    100 ml
  • Désignation:
    Trypsin, 0,05% 1X, with 0,5 g porcine trypsin in HBSS with 0,2 g/l EDTA, without calcium, magnesium (liquid)
Spécifications