Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
PIG® absorbent pillows

PIG® absorbent pillows

Pillows, PP skin/cellulose filler, 121 L, 16 pillows/box, 430×530×50 mm
N° de catalogue 121-0539
Fournisseur:   NEW PIG
PIG® absorbent pillows
N° de catalogue 121-0539
Fournisseur:  NEW PIG
PIG® absorbent pillows
PIG® absorbent pillows

Pillows, PP skin/cellulose filler, 121 L, 16 pillows/box, 430×530×50 mm

Prix
130,00 €
Connexion Pour la disponibilité
1 * 16 ST
Product Details & Documents

Add extra absorbency to your spill cleanup efforts with high-capacity PIG Pillows. Also great for steady leaks and drips.

  • Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up liquids
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Cellulose filler is highly absorbent for long-term use or big spill cleanup
  • Compact design is great for catching persistent drips and leaks in small spaces or adding absorbency for spill response
  • Absorbs and retains oils, coolants, solvents and water; not recommended for acids, caustics or other corrosive liquids

85% mixture of pre- and post-consumer recycled cellulose filler. Can be incinerated after use to reduce waste or for fuels blending.

Spécifications
  • Désignation:
    Pillows, PP skin/cellulose filler
  • Seq:
    02
  • l×p×h:
    430×530×50 mm
  • Capacité d'absorption:
    121 L
  • Emballage:
    16 pillows/box
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.