Agarose, SeaKem® HE
Agarose, SeaKem® HE
Agarose, SeaKem® HE
Catalog # 733-1556
Supplier:   LONZA
Agarose, SeaKem® HE
Catalog # 733-1556
Supplier:  LONZA
Agarose, SeaKem® HE
Agarose, SeaKem® HE
Agarose, SeaKem® HE
Price
160,00 €
Log in for availability
1 * 25 g
Product Details & Documents
SeaKem® HE agarose is a high EEO agarose that provides enhanced resolution in immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CIEP), and serum protein electrophoresis.
Specifications
  • Désignation:
    Agarose, SeaKem® HE
  • Seq:
    01