Agarose, SeaKem® HE
Agarose, SeaKem® HE
Agarose, SeaKem® HE
N° de catalogue 733-1555
Fournisseur:   LONZA
Agarose, SeaKem® HE
N° de catalogue 733-1555
Fournisseur:  LONZA
Agarose, SeaKem® HE
Agarose, SeaKem® HE
Agarose, SeaKem® HE
Prix
530,00 €
Connexion Pour la disponibilité
1 * 125 g
Product Details & Documents
SeaKem® HE agarose is a high EEO agarose that provides enhanced resolution in immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CIEP), and serum protein electrophoresis.
Spécifications
  • Désignation:
    Agarose, SeaKem® HE
  • Seq:
    02