DNA ladder
DNA ladder
100 bp ladder, 10 fragments, with 100 ng/µl concentration, 100 Assays
N° de catalogue LONZ50321
Fournisseur:   LONZA
DNA ladder
N° de catalogue LONZ50321
Fournisseur:  LONZA
Numéro CAS:  
DNA ladder
DNA ladder
100 bp ladder, 10 fragments, with 100 ng/µl concentration, 100 Assays
Numéro CAS:  
Prix
224,00 €
Connexion Pour la disponibilité
1 * 1 EA
Product Details & Documents
Thse DNA ladders provides rapid and easy size determination of the DNA samples. Each band has an ideal 50 % G-C content for precise sizing and sharp resolution. It is free from contaminating plasmid DNA with minimal background.
Spécifications
Spécifications
  • Cdt:
    100 Assays
  • Désignation:
    100 bp ladder, 10 fragments, with 100 ng/µl concentration