Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Bottle top burettes

Bottle top burettes

25 ml, ≤±0,2 %, ≤0,1 %
N° de catalogue 613-2167
Fournisseur:   VITLAB
Bottle top burettes
N° de catalogue 613-2167
Fournisseur:  VITLAB
Bottle top burettes
Bottle top burettes

25 ml, ≤±0,2 %, ≤0,1 %

Prix
1 500,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

The continuous bottle-top burette enables continuous titration, which leads to rapid, convenient, and accurate results.

  • Long operating life from two easily-replaced micro-batteries 1,5 V
  • Large, easy-to-read digits
  • Simple, media-specific readjustment via keypad
  • Continuous and precise regulation of titration speed with a large practical thumbwheel
  • Reduced titrant waste with innovative recirculation valve
  • Adaptable to bottles of different sizes
  • Telescopic discharge tube which can be adjusted in both height and length
  • Telescopic, extractable intake tube adapts burette to bottles of varying heights
  • Continual supply of titration medium with specially developed double-piston pump

The angled display shows 4-position titration volume in large, easily read numbers, which simplifies operation. Turning the
two hand wheels supplies the titration medium in a continuous and pulse-free manner via the specially developed double-piston pump. Filling procedures are not necessary. This innovative technology increases safety; its compact design and low centre of gravity reduce risk of overturning, especially with smaller bottles. The height and length of the discharge tube can be adjusted, making it possible to work safely with both short and tall bottles. The innovative recirculation system prevents the loss of valuable reagent and reduces the risk of splashes. With its simple-to-use calibration function, continuous fulfils the corresponding requirements for test equipment monitoring without instrument downtime. Margins of error are under those specified in the DIN EN ISO 8655-3 standard, even for partial volumes. VITLAB® continuous is DE-M marked. Also available with DAkkS calibration certificate.

Certifications: DIN 12600, DIN EN ISO 8655-3

Livraison: Supplied with thread adapters PP GL 32, GL 38 and S*40, a telescopic intake tube (200 to 350 mm), a telescopic discharge tube (140 to 220 mm), two micro-batteries of 1,5 volts (LR 03/AAA), and a quality certificate.

Spécifications
  • Volume:
    25 ml
  • Précision:
    ≤±0,2 %
  • Imprécision:
    ≤0,1 %
Accessories

Informations sur le produit

  • N° de catalogue
    Désignation
    Longueur
    Disponibilité
    Prix
  • VITL1671085
    Telescopic intake tube, FEP, ETFE, PTFE
    200 - 350 mm
    Connexion Pour la disponibilité
    1 * 1 ST - 56,20 €

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.