Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
10 ml, 10/19
N° de catalogue NEUB0293-04-0010
Volumetric flasks, class A, blue markings
N° de catalogue NEUB0293-04-0010
Volumetric flasks, class A, blue markings
10 ml, 10/19
Prix
23,90 €
Connexion Pour la disponibilité
1 * 1 Fl
Product Details & Documents
Borosilicate glass, grade 3.3, with PE stopper.
  • With one mark
  • Good chemical resistance
  • Good temperature resistance
Spécifications
  • RN:
    10/19
  • Capacité:
    10 ml
  • Seq:
    02