Volumetric flasks, class A, blue markings
10 ml, 10/19
N° de catalogue NEUB0293-04-0010
Fournisseur: NEUBERT VOLUME GLASSWAERE
Volumetric flasks, class A, blue markings
N° de catalogue NEUB0293-04-0010
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
Borosilicate glass, grade 3.3, with PE stopper.
- With one mark
- Good chemical resistance
- Good temperature resistance
Spécifications
- RN:10/19
- Capacité:10 ml
- Seq:02