Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Thymolphthaleine, Supelco®

Thymolphthaleine, Supelco®

Glass bottle for solids, 50 g
Catalog # SUPL89360-50G
Supplier:   Merck
Thymolphthaleine, Supelco®
Catalog # SUPL89360-50G
Supplier:  Merck
CAS Number:  125-20-2
Thymolphthaleine, Supelco®

Glass bottle for solids, 50 g

CAS Number:  125-20-2
Price
167,00 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:12,09542
  • Melting Pt:253 °C
  • Storage Temperature:Température ambiante
  • EINECS:204-729-7

Documents

Specifications

Specifications

  • Type de conditionnement:
    Glass bottle for solids
  • Cdt:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.