Operating supplies
Pump tubing, SIPS
N° de catalogue VARI9910075900
Fournisseur: VARIAN
Aller à :
Product Details & Documents
The operating supplies set includes two 500 ml constant pressure bottle, a diluent bottle (1 L), a three-way SIPS tee piece assembly for SIPS-20, six SIPS pump tubings, a SIPS kit and ten pack SIPS pump bands.
Spécifications
- Modèle:SIPS
- Désignation:Pump tubing
- Seq:02