Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Valproic Acid, Reference Standards

Valproic Acid, Reference Standards

Valproic Acid Related Compound B, USP reference standard, Ampule, 50 mg
N° de catalogue 1708718.
Fournisseur:   USP
Valproic Acid, Reference Standards
N° de catalogue 1708718.
Fournisseur:  USP
Numéro CAS:  
Valproic Acid, Reference Standards

Valproic Acid Related Compound B, USP reference standard, Ampule, 50 mg

Numéro CAS:  
Prix
1 330,00 €
Connexion Pour la disponibilité
1 * 50 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certificeringen: US Pharmacopea Reference Standard

Spécifications

Spécifications

  • Type de conditionnement:
    Ampule
  • Cdt:
    50 mg
  • Normes:
    USP reference standard
  • Désignation:
    Valproic Acid Related Compound B
  • Cat. No.:
    1708718.
  • N ° CAS.:
    62391-99-5
  • Formula:
    C₈H₁₆O₂
  • Désignation:
    Valproic Acid Related Compound B
  • Synonyme(s):
    (2RS)-2-(1-methylethyl)pentanoic acid

Spécifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.