Iron ore
NIST-693 Iron ore, nimba - constituents, Fer, Fe, 100 g
PROONIST-693
This reference standard is intended for use in checking chemical methods of analysis and in the calibration with instrumental methods of analysis.
: Traceable to NIST.
- Cdt:100 g
- Désignation:NIST-693 Iron ore, nimba - constituents
- Élement:Fer
- Symbole:Fe