Iron ore
NIST-692 Iron ore, labrador - constituents, Fer, Fe, 100 g
N° de catalogue PROONIST-692
Fournisseur: LGC Standards PROMOCHEM
Aller à :
Product Details & Documents
This reference standard is intended for use in checking chemical methods of analysis and in the calibration with instrumental methods of analysis.
Certifications: Traceable to NIST.
Spécifications
Spécifications
- Cdt:100 g
- Désignation:NIST-692 Iron ore, labrador - constituents
- Élement:Fer
- Symbole:Fe