Iron ore
Iron ore
NIST-691 Iron ore, reduced - constituents, Fer, Fe, 100 g
N° de catalogue PROONIST-691
Fournisseur:   LGC Standards PROMOCHEM
Iron ore
N° de catalogue PROONIST-691
Numéro CAS:  
Iron ore
NIST-691 Iron ore, reduced - constituents, Fer, Fe, 100 g
Numéro CAS:  
Prix
1 360,00 €
Connexion Pour la disponibilité
1 * 100 g
Product Details & Documents
This reference standard is intended for use in checking chemical methods of analysis and in the calibration with instrumental methods of analysis.
Certifications: Traceable to NIST.
Spécifications
Spécifications
  • Cdt:
    100 g
  • Désignation:
    NIST-691 Iron ore, reduced - constituents
  • Élement:
    Fer
  • Symbole:
    Fe