Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Iode 99+%, pur

Iode 99+%, pur

Flacon verre pour solides, 1 kg
Catalog # ACRO387050010
Irritant
Health
Environment
Iode 99+%, pur
Catalog # ACRO387050010
CAS Number:  7553-56-2
Iode 99+%, pur
Iode 99+%, pur

Flacon verre pour solides, 1 kg

CAS Number:  7553-56-2
Price
379,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:7553-56-2
  • Formula:I₂
  • MDL Number:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Merck Index:13,05036
  • Boiling Pt:185 °C (760 mmHg)
  • Melting Pt:113 °C
  • Density:4,9 g/cm³ (20 °C)
  • Storage Temperature:Température ambiante
  • EINECS:231-442-4
  • UN:3495
  • ADR:8,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Dark grey Metallic spheres
  • Titration Iodometric:
    ≥99.0 %
  • Insoluble matter:
    Pass

Specifications

  • Type de conditionnement:
    Flacon verre pour solides
  • Cdt:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.