Concentrator
Concentrator
Concentrateur, poudre, à utiliser avec Tube-O-DIALYZER™, 150 g
Catalog # 786-144
Supplier:   G-Biosciences
Concentrator
Catalog # 786-144
Supplier:  G-Biosciences
CAS Number:  
Concentrator
Concentrateur, poudre, à utiliser avec Tube-O-DIALYZER™, 150 g
CAS Number:  
Price
155,00 €
Log in for availability
1 * 150 g
Product Details & Documents
The concentrator is a high molecular weight polymer which will not migrate across the dialysis membrane. The concentrator is placed against the dialysis membrane of the Tube-O-DIALYZER™, where it rapidly absorbs water from the sample and reduces the sample volume. The concentrator can be used with dialysis bags and other dialysis devices.
Specifications
Specifications
  • Cdt:
    150 g
  • Désignation:
    Concentrateur, poudre, à utiliser avec Tube-O-DIALYZER™