Protein reducing agents
Protein reducing agents
OneQuant™ DTT, 0,5 M, 40
N° de catalogue 786-077
Fournisseur:   G-Biosciences
Protein reducing agents
N° de catalogue 786-077
Fournisseur:  G-Biosciences
Numéro CAS:  
Protein reducing agents
Protein reducing agents
OneQuant™ DTT, 0,5 M, 40
Numéro CAS:  
Prix
253,00 €
Connexion Pour la disponibilité
1 * 40 ST
Product Details & Documents
Protein-S-S-Reductant™ uses TCEP (Tris [2-carboxyethyl] phosphine), a popular alternative to β-mercaptoethanol and DTT (dithiothreitol). TCEP improves stability, increases effectiveness, and reduces proteins over a wider range of pH, including lower acidic pHs. Protein-S-S-Reductant™ completely reduces stable disulfide bonds in less than 5 minutes at room temperature and is compatible with the protein alkylation reactions. This ready to use solution is at a neutral pH and stabilised for long-term storage. Simply supplement Protein-S-S-Reductant™ in place of DTT or β-mercaptoethanol and boil the sample.
OneQuant™ DTT are single aliquots of DTT that eliminate the need for weighing; preventing loss of reagent and saving time. Add 90 μl water to a single tube to generate a 0,5 M DTT solution. Supplied with 40 individual tubes.
Spécifications
Spécifications
  • Cdt:
    40
  • Désignation:
    OneQuant™ DTT, 0,5 M