DNAOUT™ DNA Removal Solution
DNAOUT™ DNA Removal Solution
DNA OUT™, refill, 1 L
N° de catalogue 786-75
Fournisseur:   G-Biosciences
DNAOUT™ DNA Removal Solution
N° de catalogue 786-75
Fournisseur:  G-Biosciences
Numéro CAS:  
DNAOUT™ DNA Removal Solution
DNAOUT™ DNA Removal Solution
DNA OUT™, refill, 1 L
Numéro CAS:  
Prix
113,00 €
Connexion Pour la disponibilité
1 * 1 L
Product Details & Documents
DNAOUT™ is a DNA removal solution for the rapid decontamination of work areas from contaminating DNA. Simply spray the work area and wait 10-15 seconds and then rinse with Molecular Grade Water.
  • Ready-to-use with convenient spray bottle and refill bottles available
  • Remove contaminating DNA prior to sensitive work, including PCR
  • Removes amplicon, plasmid and genomic DNA
  • Fast acting (10-15 seconds)
  • Ideal for cleaning centrifuge tubes, reaction tubes, PCR machine surface, pipettes, lab benches, lab equipment and more
Spécifications
Spécifications
  • Cdt:
    1 L
  • Désignation:
    DNA OUT™, refill
  • Cat. No.:
    786-75