Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Eau, BAKER ANALYZED®, J.T.Baker®

Eau, BAKER ANALYZED®, J.T.Baker®

Flacon Verre, 1 L
N° de catalogue 9825.1000GL
   Safer Human and Environmental Health
Fournisseur:   Avantor
Eau, BAKER ANALYZED®, J.T.Baker®
N° de catalogue 9825.1000GL
Fournisseur:  Avantor
   Safer Human and Environmental Health
Numéro CAS:  7732-18-5
Eau, BAKER ANALYZED®, J.T.Baker®

Flacon Verre, 1 L

Numéro CAS:  7732-18-5
Prix
49,25 €
Connexion Pour la disponibilité
1 * 1 L
Product Details & Documents
Synonymes:  Hydrogen oxide
  • Numéro CAS:7732-18-5
  • Formule:H₂O
  • Numéro MDL:MFCD00011332
  • Poids moléculaire:18.02 g/mol
  • Index Merck:13,10098
  • Point d'ébullition:100 °C (1013 hPa)
  • Point de fusion:0 °C
  • Densité:1 g/cm³ (4 °C)
  • Température de stockage:Température ambiante
  • EINECS:231-791-2
Spécifications

Résultats des tests de spécification

  • Residue after Evaporation:
    max. 5 ppm
  • LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid::
    -
  • at 210 nm:
    max. 0,005
  • at 254 nm:
    max. 0,001 uv
  • LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid::
    -
  • Positive ESI-MS Sensitive Impurities (as Reserpine):
    max. 100 ng/ml
  • Trace Impurities (in ppb):
    -
  • Aluminium (Al):
    max. 500 ppb
  • Calcium (Ca):
    max. 100 ppb
  • Iron (Fe):
    max. 100 ppb
  • Magnesium (Mg):
    max. 100 ppb
  • Potassium (K):
    max. 100 ppb
  • Sodium (Na):
    max. 100 ppb
  • Product Information (not specifications)::
    -
  • Density (g/ml) at 20°C:
    1.00

Spécifications

  • Type de conditionnement:
    Flacon Verre
  • Cdt:
    1 L
  • Cat. No.:
    9825.1000GL
  • Certification environnementale:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.