Drying tubes, U-shaped
Drying tubes, U-shaped
19/26 NS, 150 mm, 18 mm
N° de catalogue WITG2.114.150
Fournisseur:   witeg Labortechnik
Drying tubes, U-shaped
N° de catalogue WITG2.114.150
Fournisseur:  witeg Labortechnik
Drying tubes, U-shaped
Drying tubes, U-shaped
19/26 NS, 150 mm, 18 mm
Prix
70,30 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Spécifications
  • Ø int.:
    18 mm
  • Rodage femelle:
    19/26 NS
  • Longueur:
    150 mm