Accessories for cold traps
Cold trap SL 29 GL-A, spare part, with spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml
Catalog # KGWA17387
Supplier:   KGW
Accessories for cold traps
Catalog # KGWA17387
Supplier:  KGW
Accessories for cold traps
Cold trap SL 29 GL-A, spare part, with spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml
Price
347,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Désignation:
    Cold trap SL 29 GL-A, spare part, with spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml
  • Seq:
    08