Cold traps, Type KF
Cold trap S 29 GL-A, borosilicate glass 3,3, with threaded glass joint GL 18 and PTFE olive, 150 ml condensate capacity
Catalog # KGWA17377
Supplier: KGW
Jump to:
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
- Easy to clean
- No stand material for holding the cold trap
- Protective casing of Dewar flask made of blue coated metal or aluminium
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : [email protected]
Specifications
- Désignation:Cold trap S 29 GL-A, borosilicate glass 3,3, with threaded glass joint GL 18 and PTFE olive, 150 ml condensate capacity
- Seq:05