DNA ladder
DNA ladder
500 bp ladder, 16 fragments, with 200 ng/µl concentration, 100 Tests
Catalog # LONZ50323
Supplier:   LONZA
DNA ladder
Catalog # LONZ50323
Supplier:  LONZA
CAS Number:  
DNA ladder
DNA ladder
500 bp ladder, 16 fragments, with 200 ng/µl concentration, 100 Tests
CAS Number:  
Price
224,00 €
Log in for availability
1 * 1 EA
Product Details & Documents
Thse DNA ladders provides rapid and easy size determination of the DNA samples. Each band has an ideal 50 % G-C content for precise sizing and sharp resolution. It is free from contaminating plasmid DNA with minimal background.
Specifications
Specifications
  • Cdt:
    100 Tests
  • Désignation:
    500 bp ladder, 16 fragments, with 200 ng/µl concentration