Sample spoons, mini, SP Bel-Art
Sample spoons, mini, SP Bel-Art
0,05 ml, PS
N° de catalogue BELAF367210005
Sample spoons, mini, SP Bel-Art
N° de catalogue BELAF367210005
Sample spoons, mini, SP Bel-Art
Sample spoons, mini, SP Bel-Art
0,05 ml, PS
Prix
56,70 €
Connexion Pour la disponibilité
1 * 25 ST
Product Details & Documents
PS, white.
  • Disposable, made from white high-impact PS
  • Ideal for collecting specific small volumes of powders and other dry materials
  • 0,10 / 0,50 / 1,0 ml models have a rounded bowl for easier emptying of the spoon and a thinner neck that minimises extra material collection
Documents
Spécifications
  • Capacité:
    0,05 ml
  • Matière:
    PS
  • Seq:
    01