Adapters with stopcock and socket
Adapters with stopcock and socket
19/26 NS, 3 mm
N° de catalogue 201-3019
Fournisseur:   SciLabware
Adapters with stopcock and socket
N° de catalogue 201-3019
Fournisseur:  SciLabware
Adapters with stopcock and socket
Adapters with stopcock and socket
19/26 NS, 3 mm
Prix
115,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With GP Rotaflo® PTFE stopcock providing fine flow control and positive closure. Detachable plastic screwthread connectors allow rubber hose etc. to be fitted easily and safely .
Spécifications
  • Ø trou:
    3 mm
  • Rodage femelle:
    19/26 NS