Adapters with stopcock and socket
19/26 NS, 3 mm
N° de catalogue 201-3019
Fournisseur: SciLabware
Aller à :
Product Details & Documents
PYREX®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With GP Rotaflo® PTFE stopcock providing fine flow control and positive closure. Detachable plastic screwthread connectors allow rubber hose etc. to be fitted easily and safely .
Spécifications
- Ø trou:3 mm
- Rodage femelle:19/26 NS