NMR sample tubes, Standard series
NMR sample tubes, Standard series
203 mm, ± 13 µm, ± 5 µm, MHz: 500
N° de catalogue 637-0199
NMR sample tubes, Standard series
N° de catalogue 637-0199
NMR sample tubes, Standard series
NMR sample tubes, Standard series
203 mm, ± 13 µm, ± 5 µm, MHz: 500
Prix
101,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Glass, Class A, ASTM Type 1.
Spécifications
  • Courbures:
    ± 13 µm
  • Concentricité:
    ± 5 µm
  • Type:
    MHz: 500
  • Longueur:
    203 mm
  • Seq:
    05