Dressing jars
Dressing jars
Lids, moulded glass, circular, for jars of O.Ø 125 mm
N° de catalogue MURRG501/02
Fournisseur:   MURRAY S
Dressing jars
N° de catalogue MURRG501/02
Fournisseur:  MURRAY S
Dressing jars
Lids, moulded glass, circular, for jars of O.Ø 125 mm
Prix
28,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Dressing jars, made of moulded glass, round.
  • Good chemical resistance
  • Lids must be ordered separately
Spécifications
  • Désignation:
    Lids, moulded glass, circular, for jars of O.Ø 125 mm
  • Seq:
    12