Test tubes, without rim, light wall
150 mm, 18 mm, 0,8 mm
N° de catalogue MURRG021/37
Fournisseur: MURRAY S
Aller à :
Product Details & Documents
Glass, clear.
- Excellent chemical resistance
- Good temperature resistance
Spécifications
- Ø ext.:18 mm
- Épaisseur de paroi:0,8 mm
- Longueur:150 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



